Assay Details
Target Gene Details
Entrez Gene ID: | 13404 |
Gene Name: | DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologous recombination |
Gene Aliases: |
Dmc1h, Lim15, Mei11, sgdp |
Location: |
Chr.15:79561497-79605154 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmc1 | NM_001278226.1 | NP_001265155.1 | ||
| NM_010059.3 | NP_034189.1 | |||
| NR_103477.1 | ||||
| AK076777.1 | BAC36477.1 | |||
| AK169570.1 | ||||
| BC116767.1 | ||||
| BC119081.1 | ||||
| D58419.1 | BAA09590.1 | |||
| D64107.1 | BAA10969.1 |
More Information
Set Membership: |
|

Genomic Map