Assay Details
Target Gene Details
Entrez Gene ID: | 252973 |
Gene Name: | grainyhead-like 2 (Drosophila) |
Gene Aliases: |
0610015A08Rik, BOM, Tcfcp2l3 |
Location: |
Chr.15:37233036-37363569 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grhl2 | NM_026496.4 | NP_080772.2 | ||
| AF411213.1 | AAM22619.1 | |||
| AK002633.1 | BAB22247.1 | |||
| BC055035.1 | AAH55035.1 | |||
| BY703140.1 |
More Information
Set Membership: |
|

Genomic Map