Assay Details
Target Gene Details
Entrez Gene ID: | 15382 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A1 |
Gene Aliases: |
HDP-1, Hdp, Hnrpa1, hnRNP A1, hnrnp-A1 |
Location: |
Chr.15:103240397-103246698 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |

Genomic Map