Assay Details
Target Gene Details
Entrez Gene ID: | 268782 |
Gene Name: | alanine-glyoxylate aminotransferase 2 |
Gene Aliases: |
AI303810, AI663818 |
Location: |
Chr.15:10358516-10410159 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agxt2 | NM_001031851.1 | NP_001027021.1 | ||
| AK149538.1 | BAE28945.1 | |||
| BC141248.1 | ||||
| BC145576.1 |

Genomic Map