Assay Details
Target Gene Details
Entrez Gene ID: | 668218 |
Gene Name: | bridging integrator 2 |
Gene Aliases: |
- |
Location: |
Chr.15:100641082-100669576 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bin2 | NM_001270537.1 | NP_001257466.1 | ||
| CN691011.1 | ||||
| DV661115.1 |
Target Gene Details
Entrez Gene ID: | 102465580 |
Gene Name: | microRNA 6961 |
Gene Aliases: |
Gm27367, mmu-mir-6961 |
Location: |
Chr.15:100649163-100649229 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir6961 | NR_105926.1 |

Genomic Map