Assay Details
Target Gene Details
Entrez Gene ID: | 13132 |
Gene Name: | disabled 2, mitogen-responsive phosphoprotein |
Gene Aliases: |
5730435J12Rik, AA960054, AI957090, D15Wsu122e, D630005B22Rik, Doc-2, Doc2, p96 |
Location: |
Chr.15:6299788-6440715 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dab2 | NM_001008702.2 | NP_001008702.1 | ||
| NM_001037905.3 | NP_001032994.1 | |||
| NM_001102400.1 | NP_001095870.1 | |||
| NM_023118.5 | NP_075607.2 | |||
| AK154716.1 | BAE32784.1 | |||
| AK162744.1 | BAE37047.1 | |||
| AK162851.1 | BAE37084.1 | |||
| AK165519.1 | BAE38234.1 | |||
| AK165810.1 | BAE38389.1 | |||
| AK168375.1 | ||||
| BC006588.1 | AAH06588.1 | |||
| BC016887.1 | AAH16887.1 | |||
| U18869.1 | AAB02646.1 |
More Information
Set Membership: |
|

Genomic Map