Assay Details
Target Gene Details
Entrez Gene ID: | 15382 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A1 |
Gene Aliases: |
HDP-1, Hdp, Hnrpa1, hnRNP A1, hnrnp-A1 |
Location: |
Chr.15:103240397-103246698 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpa1 | NM_001039129.4 | NP_001034218.1 | ||
| NM_010447.5 | NP_034577.1 | |||
| NR_104427.1 | ||||
| AK007802.1 | BAB25267.1 | |||
| AK088308.1 | BAC40273.1 | |||
| AK131999.1 | BAE20929.1 | |||
| AK135391.1 | BAE22518.1 | |||
| AK136113.1 | ||||
| AK146126.1 | BAE26919.1 | |||
| AK146630.1 | BAE27318.1 | |||
| AK152684.1 | BAE31416.1 | |||
| AK161730.1 | BAE36552.1 | |||
| AK166901.1 | BAE39104.1 | |||
| AK166989.1 | BAE39169.1 | |||
| AK167161.1 | BAE39302.1 | |||
| AK167810.1 | BAE39837.1 | |||
| AK167811.1 | BAE39838.1 | |||
| AK167820.1 | ||||
| AK167913.1 | BAE39920.1 | |||
| AK168420.1 | BAE40333.1 | |||
| AK168531.1 | BAE40409.1 | |||
| AK168733.1 | BAE40574.1 | |||
| AK169216.1 | BAE40987.1 | |||
| BC080675.1 | AAH80675.1 | |||
| BC083136.1 | AAH83136.1 | |||
| BC089340.1 | AAH89340.1 | |||
| BC092395.1 | AAH92395.1 | |||
| D86728.1 | BAA13161.1 | |||
| D86729.1 | BAA13162.1 | |||
| M99167.1 | AAA37633.1 |

Genomic Map