Assay Details
Target Gene Details
Entrez Gene ID: | 239447 |
Gene Name: | collectin sub-family member 10 |
Gene Aliases: |
CL-L1 |
Location: |
Chr.15:54410774-54466360 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Colec10 | NM_173422.3 | NP_775598.2 | ||
| AB016429.1 | BAC53954.1 | |||
| AK028423.1 | BAC25941.1 | |||
| BC106836.1 | AAI06837.1 | |||
| BC106837.1 | AAI06838.1 | |||
| BC108945.1 | AAI08946.1 |
More Information
Set Membership: |
|

Genomic Map