Assay Details
Target Gene Details
Entrez Gene ID: | 27373 |
Gene Name: | casein kinase 1, epsilon |
Gene Aliases: |
AI426939, AI551861, AW457082, CK1epsilon, CKIe, KC1epsilon, tau |
Location: |
Chr.15:79417852-79455579 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csnk1e | NM_001289898.1 | NP_001276827.1 | ||
| NM_001289899.1 | NP_001276828.1 | |||
| NM_013767.6 | NP_038795.3 | |||
| AB028241.1 | BAA88082.1 | |||
| AB028736.2 | BAA88107.2 | |||
| AK158709.1 | BAE34622.1 | |||
| BC026127.1 | AAH26127.1 |
More Information
Set Membership: |
|

Genomic Map