Assay Details
Target Gene Details
Entrez Gene ID: | 15499 |
Gene Name: | heat shock factor 1 |
Gene Aliases: |
AA960185, Hsf1alpha, Hsf1beta |
Location: |
Chr.15:76477395-76500978 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsf1 | NM_008296.2 | NP_032322.1 | ||
| AK075651.1 | ||||
| AK089144.1 | ||||
| BC013716.1 | AAH13716.1 | |||
| BC094064.1 | AAH94064.1 | |||
| KM043052.1 | ||||
| KM043053.1 | ||||
| KM043054.1 | ||||
| KM043055.1 | ||||
| X61753.1 | CAA43892.1 |
More Information
Set Membership: |
|

Genomic Map