Assay Details
Target Gene Details
Entrez Gene ID: | 78541 |
Gene Name: | ankyrin repeat and SOCS box-containing 8 |
Gene Aliases: |
4930539L19Rik, AI788835, C430011H06Rik |
Location: |
Chr.15:98134637-98165602 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb8 | NM_001170710.1 | NP_001164181.1 | ||
| NM_001170711.1 | NP_001164182.1 | |||
| NM_030121.4 | NP_084397.3 | |||
| AF398969.1 | AAK97491.1 | |||
| AK016001.1 | BAB30077.1 | |||
| AK021239.1 | ||||
| AK030168.1 | BAC26819.1 | |||
| AK087299.1 | ||||
| AK182854.1 | ||||
| AK214224.1 | ||||
| BC025106.1 | AAH25106.1 | |||
| BY240190.1 |

Genomic Map