Assay Details
Target Gene Details
Entrez Gene ID: | 70472 |
Gene Name: | ATPase family, AAA domain containing 2 |
Gene Aliases: |
2610509G12Rik |
Location: |
Chr.15:58094047-58135139 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atad2 | NM_027435.2 | 11 | 1374 | NP_081711.2 |
| AK029867.1 | 4 | 521 | BAC26651.1 | |
| AK032877.1 | 11 | 1375 |
More Information
Set Membership: |
|

Genomic Map