Assay Details
Target Gene Details
Entrez Gene ID: | 16197 |
Gene Name: | interleukin 7 receptor |
Gene Aliases: |
CD127, IL-7Ralpha |
Location: |
Chr.15:9506159-9529876 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il7r | NM_008372.4 | 3 | 444 | NP_032398.3 |
| AF078906.1 | 3 | 310 | AAF06717.1 | |
| AK040364.1 | 3 | 427 | BAC30571.1 | |
| AK040740.1 | 3 | 427 | BAC30689.1 | |
| AK041838.1 | 3 | 335 | BAC31080.1 | |
| AK042264.1 | 3 | 389 | BAC31208.1 | |
| AK154215.1 | 2 | 258 | ||
| AK154516.1 | 3 | 404 | BAE32645.1 | |
| BC089571.1 | 3 | 396 | AAH89571.1 | |
| M29697.1 | 3 | 357 | AAA39304.1 |
More Information
Set Membership: |
|

Genomic Map