Assay Details
Target Gene Details
Entrez Gene ID: | 16197 |
Gene Name: | interleukin 7 receptor |
Gene Aliases: |
CD127, IL-7Ralpha |
Location: |
Chr.15:9506159-9529876 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il7r | NM_008372.4 | NP_032398.3 | ||
| AF078906.1 | AAF06717.1 | |||
| AK040364.1 | BAC30571.1 | |||
| AK042264.1 | BAC31208.1 | |||
| AK154215.1 | ||||
| AK154516.1 | BAE32645.1 | |||
| BC089571.1 | AAH89571.1 | |||
| M29697.1 | AAA39304.1 |
More Information
Set Membership: |
|

Genomic Map