Assay Details
Target Gene Details
Entrez Gene ID: | 11600 |
Gene Name: | angiopoietin 1 |
Gene Aliases: |
1110046O21Rik, Ang-1, Ang1 |
Location: |
Chr.15:42424667-42676977 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Angpt1 | NM_001286062.1 | NP_001272991.1 | ||
| NM_009640.4 | NP_033770.2 | |||
| AK088439.1 | BAC40354.1 | |||
| AK172868.1 | BAD32146.1 | |||
| BC067410.1 | AAH67410.1 | |||
| U83509.1 | AAB50558.1 |
More Information
Set Membership: |
|

Genomic Map