Assay Details
Target Gene Details
Entrez Gene ID: | 11908 |
Gene Name: | activating transcription factor 1 |
Gene Aliases: |
- |
Location: |
Chr.15:100227859-100261248 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atf1 | NM_007497.3 | NP_031523.3 | ||
| AK010428.1 | BAB26930.1 | |||
| AK144511.1 | ||||
| BC006871.1 | AAH06871.1 | |||
| BY733806.1 | ||||
| M63725.1 | AAA40395.1 |

Genomic Map