Assay Details
Target Gene Details
Entrez Gene ID: | 66445 |
Gene Name: | cytochrome c-1 |
Gene Aliases: |
2610002H19Rik, AA408921 |
Location: |
Chr.15:76343523-76345934 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyc1 | NM_025567.2 | NP_079843.1 | ||
| AK002815.1 | BAB22380.1 | |||
| AK011288.1 | BAB27518.1 | |||
| AK151299.1 | BAE30282.1 | |||
| AK170272.1 | BAE41677.1 | |||
| AK182781.1 | ||||
| AK184866.1 | ||||
| AK188690.1 | ||||
| AK192516.1 | ||||
| AK195664.1 | ||||
| AK215816.1 | ||||
| BC005620.1 | AAH05620.1 | |||
| BC082790.1 | AAH82790.1 |

Genomic Map