Assay Details
Target Gene Details
Entrez Gene ID: | 406223 |
Gene Name: | predicted gene 5414 |
Gene Aliases: |
EG406223, Kb14 |
Location: |
Chr.15:101624028-101628188 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5414 | NM_001003670.1 | NP_001003670.1 |

Genomic Map