Assay Details
Target Gene Details
Entrez Gene ID: | 432982 |
Gene Name: | predicted gene 5475 |
Gene Aliases: |
EG432982 |
Location: |
Chr.15:100423193-100428150 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5475 | NR_040351.1 | |||
| AK163173.1 | BAE37220.1 | |||
| BC147350.1 | ||||
| BC147351.1 |
More Information
Set Membership: |
|

Genomic Map