Assay Details
Target Gene Details
Entrez Gene ID: | 54151 |
Gene Name: | cysteine and histidine rich 1 |
Gene Aliases: |
1110031M01Rik, AU042374, Chrp |
Location: |
Chr.15:76643395-76660208 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyhr1 | NM_001276321.1 | 2 | 662 | NP_001263250.1 |
| NM_019396.3 | 3 | 595 | NP_062269.1 | |
| AJ251516.1 | 3 | 448 | CAB62575.1 | |
| AK080867.1 | 2 | 682 | ||
| AK129156.1 | 1 | 1769 | BAC97966.1 | |
| BC062184.1 | 2 | 651 | AAH62184.1 | |
| BC082772.1 | 2 | 662 | AAH82772.1 | |
| BE653118.1 | ||||
| BX512375.1 | 2 | 201 | ||
| BY287678.1 | 2 | 226 | ||
| CF742880.1 | 1 | 110 |

Genomic Map