Assay Details
Target Gene Details
Entrez Gene ID: | 16402 |
Gene Name: | integrin alpha 5 (fibronectin receptor alpha) |
Gene Aliases: |
Cd49e, Fnra, VLA5 |
Location: |
Chr.15:103344286-103366748 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itga5 | NM_010577.3 | NP_034707.3 | ||
| AK142241.1 | ||||
| AK144921.1 | ||||
| AK148138.1 | ||||
| AK190883.1 | ||||
| AK205630.1 | ||||
| BC050943.1 | AAH50943.1 | |||
| BC059829.1 | AAH59829.1 | |||
| X15203.1 | CAA33273.1 | |||
| X79003.1 | CAA55638.1 |
Target Gene Details
Entrez Gene ID: | 102466775 |
Gene Name: | microRNA 6963 |
Gene Aliases: |
Gm27724, mmu-mir-6963 |
Location: |
Chr.15:103350455-103350525 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir6963 | NR_105928.1 |
More Information
Set Membership: |
|

Genomic Map