Assay Details
Target Gene Details
Entrez Gene ID: | 94230 |
Gene Name: | cleavage and polyadenylation specific factor 1 |
Gene Aliases: |
- |
Location: |
Chr.15:76595808-76607613 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpsf1 | NM_001164173.1 | NP_001157645.1 | ||
| NM_053193.2 | NP_444423.1 | |||
| AF322193.1 | AAG40326.1 | |||
| AK149914.1 | ||||
| AK155648.1 | BAE33365.1 | |||
| AK155806.1 | ||||
| BC056388.1 | AAH56388.1 |
More Information
Set Membership: |
|

Genomic Map