Assay Details
Target Gene Details
Entrez Gene ID: | 69440 |
Gene Name: | DENN/MADD domain containing 6B |
Gene Aliases: |
1700027J05Rik, AI414881, Fam116b |
Location: |
Chr.15:89182213-89196474 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd6b | NM_027081.3 | 4 | 398 | NP_081357.2 |
| AK084555.1 | 4 | 391 | ||
| AK154949.1 | 4 | 420 | ||
| BB658928.1 | 4 | 391 | ||
| BC139003.1 | 4 | 372 | ||
| BY252285.1 | 4 | 401 | ||
| BY748566.1 | 4 | 415 |

Genomic Map