Assay Details
Target Gene Details
Entrez Gene ID: | 69787 |
Gene Name: | annexin A13 |
Gene Aliases: |
1810034H17Rik, AV055219 |
Location: |
Chr.15:58341464-58389274 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anxa13 | NM_027211.2 | NP_081487.1 | ||
| AJ306451.1 | CAC34623.1 | |||
| AK007692.1 | BAB25193.1 | |||
| AV289596.2 | ||||
| BC013521.1 | AAH13521.1 |
More Information
Set Membership: |
|

Genomic Map