Assay Details
Target Gene Details
Entrez Gene ID: | 239556 |
Gene Name: | calcium channel, voltage-dependent, alpha 1I subunit |
Gene Aliases: |
Ca(V)3.3 |
Location: |
Chr.15:80287238-80398292 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna1i | NM_001044308.2 | NP_001037773.2 | ||
| AK147585.1 |
More Information
Set Membership: |
|

Genomic Map