Assay Details
Target Gene Details
Entrez Gene ID: | 100134861 |
Gene Name: | Chkb-Cpt1b readthrough transcript (NMD candidate) |
Gene Aliases: |
BC090627, Chkb-Cpt1b, Chkbcpt1b |
Location: |
Chr.15:89416405-89429927 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ChkbCpt1b | NR_004843.2 |
Target Gene Details
Entrez Gene ID: | 12895 |
Gene Name: | carnitine palmitoyltransferase 1b, muscle |
Gene Aliases: |
Cpt1, Cpt1-m, Cpti, Cpti-m, M-cpti |
Location: |
Chr.15:89416405-89425862 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpt1b | NM_009948.2 | NP_034078.2 | ||
| AB010826.1 | BAA88685.1 | |||
| AF017174.2 | AAC31640.2 | |||
| AK029141.1 | ||||
| AK146851.1 | BAE27481.1 | |||
| BC018270.1 | AAH18270.1 |
More Information
Set Membership: |
|

Genomic Map