Assay Details
Target Gene Details
Entrez Gene ID: | 13101 |
Gene Name: | cytochrome P450, family 2, subfamily d, polypeptide 10 |
Gene Aliases: |
AI303445, Cyp2d, P450-2D |
Location: |
Chr.15:82402845-82407208 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2d10 | NM_010005.3 | NP_034135.2 | ||
| BC010989.1 | AAH10989.1 | |||
| BC057924.1 | AAH57924.1 | |||
| BF660186.1 | ||||
| CB951489.1 | ||||
| M27167.1 | AAA39878.1 |
More Information
Set Membership: |
|

Genomic Map