Assay Details
Target Gene Details
Entrez Gene ID: | 78244 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 21 |
Gene Aliases: |
4930461P20Rik, 9930116P15Rik |
Location: |
Chr.15:10446763-10470516 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc21 | NM_030046.2 | 6 | 972 | NP_084322.2 |
| AK019636.1 | 1 | 67 | ||
| AK141303.1 | 3 | 432 | BAE24641.1 |
More Information
Set Membership: |
|

Genomic Map