Assay Details
Target Gene Details
Entrez Gene ID: | 64705 |
Gene Name: | dihydropyrimidinase |
Gene Aliases: |
1200017I10Rik, 1300004I01Rik, DHP, DHPase |
Location: |
Chr.15:39768485-39857470 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpys | NM_001164466.1 | 2 | 416 | NP_001157938.1 |
| NM_022722.3 | 2 | 416 | NP_073559.3 | |
| AF227731.1 | 2 | 334 | AAK00644.1 | |
| AF249296.1 | 2 | 403 | AAG37999.1 | |
| AK004822.1 | 2 | 416 | BAB23593.1 | |
| AK004899.2 | 2 | 413 | BAB23654.1 | |
| BC037086.1 | 2 | 385 | AAH37086.1 |
More Information
Set Membership: |
|

Genomic Map