Assay Details
Target Gene Details
Entrez Gene ID: | 16678 |
Gene Name: | keratin 1 |
Gene Aliases: |
Krt-2.1, Krt2-1, Krt86 |
Location: |
Chr.15:101845426-101850786 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt1 | NM_008473.2 | 2 | 792 | NP_032499.2 |
| AK019521.1 | 2 | 793 | BAB31776.1 | |
| BC117842.1 | 2 | 759 | ||
| BC117843.1 | 2 | 759 | ||
| M10937.1 | 2 | 800 | AAD05191.1 |

Genomic Map