Assay Details
Target Gene Details
Entrez Gene ID: | 16681 |
Gene Name: | keratin 2 |
Gene Aliases: |
BB005427, Krt2-17, Krt2-2, Krt2e |
Location: |
Chr.15:101810689-101818169 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt2 | NM_010668.2 | 2 | 879 | NP_034798.2 |
| AK132476.1 | 2 | 881 | BAE21186.1 | |
| AK161078.1 | 2 | 878 | BAE36187.1 | |
| AK161098.1 | 2 | 881 | BAE36194.1 | |
| BC120485.1 | 2 | 845 | ||
| BC139485.1 | 2 | 845 | ||
| X74784.1 | 2 | 845 | CAA52788.1 |

Genomic Map