Assay Details
Target Gene Details
Entrez Gene ID: | 328561 |
Gene Name: | apolipoprotein L 10B |
Gene Aliases: |
9130218O11Rik |
Location: |
Chr.15:77584106-77598542 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apol10b | NM_177820.4 | NP_808488.2 | ||
| AK144113.1 |
More Information
Set Membership: |
|

Genomic Map