Assay Details
Target Gene Details
Entrez Gene ID: | 380997 |
Gene Name: | cytochrome P450, family 2, subfamily d, polypeptide 12 |
Gene Aliases: |
9030605E09Rik |
Location: |
Chr.15:82554960-82559413 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2d12 | NM_201360.1 | NP_958748.1 | ||
| AK078880.1 | BAC37440.1 | |||
| BC139169.1 | ||||
| BC139170.1 | ||||
| BC145567.1 |
More Information
Set Membership: |
|

Genomic Map