Assay Details
Target Gene Details
Entrez Gene ID: | 76279 |
Gene Name: | cytochrome P450, family 2, subfamily d, polypeptide 26 |
Gene Aliases: |
1300006E06Rik |
Location: |
Chr.15:82790107-82794452 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2d26 | NM_029562.2 | NP_083838.1 | ||
| AK004915.1 | BAB23666.1 | |||
| BC023241.1 | AAH23241.1 |
More Information
Set Membership: |
|

Genomic Map