Assay Details
Target Gene Details
Entrez Gene ID: | 66270 |
Gene Name: | family with sequence similarity 134, member B |
Gene Aliases: |
1810015C04Rik, AU015349 |
Location: |
Chr.15:25843298-25973696 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam134b | NM_001034851.2 | NP_001030023.1 | ||
| NM_001277315.1 | 1 | 175 | NP_001264244.1 | |
| NM_001277316.1 | 1 | 51 | NP_001264245.1 | |
| NM_001277318.1 | NP_001264247.1 | |||
| NM_025459.3 | 1 | 51 | NP_079735.2 | |
| AK015182.1 | ||||
| AK146972.1 | 1 | 52 | BAE27577.1 | |
| AK167327.1 | 1 | 176 | BAE39429.1 | |
| AK195437.1 | ||||
| BC019494.1 | AAH19494.1 | |||
| BC054415.1 | AAH54415.1 | |||
| BY779369.1 | ||||
| CB724233.1 |

Genomic Map