Assay Details
Target Gene Details
Entrez Gene ID: | 16682 |
Gene Name: | keratin 4 |
Gene Aliases: |
AW108092, K4, Krt-2.4, Krt2-4 |
Location: |
Chr.15:101918535-101924735 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt4 | NM_008475.2 | NP_032501.2 | ||
| BC064008.1 | AAH64008.1 | |||
| BC145839.1 | ||||
| BC145841.1 | ||||
| X03491.1 | CAA27207.1 |

Genomic Map