Assay Details
Target Gene Details
Entrez Gene ID: | 118452 |
Gene Name: | brain and acute leukemia, cytoplasmic |
Gene Aliases: |
2810457D07Rik |
Location: |
Chr.15:38933909-38952917 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baalc | NM_080640.5 | NP_542371.1 | ||
| AK045383.1 | ||||
| AK079337.1 | BAC37611.1 |

Genomic Map