Assay Details
Target Gene Details
Entrez Gene ID: | 223706 |
Gene Name: | cytochrome P450, family 2, subfamily d, polypeptide 34 |
Gene Aliases: |
- |
Location: |
Chr.15:82615965-82620907 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2d34 | NM_145474.2 | NP_663449.2 |
More Information
Set Membership: |
|

Genomic Map