Assay Details
Target Gene Details
Entrez Gene ID: | 66270 |
Gene Name: | family with sequence similarity 134, member B |
Gene Aliases: |
1810015C04Rik, AU015349 |
Location: |
Chr.15:25843298-25973696 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam134b | NM_001034851.2 | 6 | 729 | NP_001030023.1 |
| NM_001277315.1 | 4 | 818 | NP_001264244.1 | |
| NM_001277316.1 | 4 | 401 | NP_001264245.1 | |
| NM_001277317.1 | 4 | 379 | NP_001264246.1 | |
| NM_001277318.1 | 4 | 351 | NP_001264247.1 | |
| NM_025459.3 | 5 | 519 | NP_079735.2 | |
| AK007506.1 | 4 | 380 | BAB25077.1 | |
| AK015182.1 | 5 | 341 | ||
| AK146972.1 | 4 | 402 | BAE27577.1 | |
| AK167327.1 | 4 | 819 | BAE39429.1 | |
| AK195437.1 | ||||
| BC019494.1 | 5 | 458 | AAH19494.1 | |
| BC054415.1 | 4 | 338 | AAH54415.1 |

Genomic Map