Assay Details
Target Gene Details
Entrez Gene ID: | 110862 |
Gene Name: | potassium voltage-gated channel, subfamily Q, member 3 |
Gene Aliases: |
- |
Location: |
Chr.15:65992662-66286813 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnq3 | NM_152923.2 | NP_690887.2 | ||
| AY118171.1 | AAM81579.1 | |||
| BC116306.1 |
More Information
Set Membership: |
|

Genomic Map