Assay Details
Target Gene Details
Entrez Gene ID: | 16687 |
Gene Name: | keratin 6A |
Gene Aliases: |
2310016L08Rik, Krt2-6a, Krt2-6c, MK6a, mK6[a] |
Location: |
Chr.15:101689928-101694305 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt6a | NM_008476.3 | NP_032502.3 | ||
| AK009394.1 | ||||
| BC080820.1 | AAH80820.1 | |||
| K02108.1 | AAA39395.1 |

Genomic Map