Assay Details
Target Gene Details
Entrez Gene ID: | 12297 |
Gene Name: | calcium channel, voltage-dependent, beta 3 subunit |
Gene Aliases: |
Beta3, CAB3, Ca(v)beta3, Cchb3 |
Location: |
Chr.15:98626873-98644536 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacnb3 | NM_001044741.2 | NP_001038206.1 | ||
| NM_001286226.1 | NP_001273155.1 | |||
| NM_007581.3 | NP_031607.2 | |||
| AF498245.1 | AAM22963.1 | |||
| AF498246.1 | ||||
| AK148088.1 | ||||
| BU709207.1 | ||||
| CD807951.1 | ||||
| CJ131971.1 | ||||
| U20372.1 | AAA91105.1 |
More Information
Set Membership: |
|

Genomic Map