Assay Details
Target Gene Details
Entrez Gene ID: | 406222 |
Gene Name: | keratin 74 |
Gene Aliases: |
K74, Kb37 |
Location: |
Chr.15:101754259-101763504 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt74 | NR_033444.1 |

Genomic Map