Assay Details
Target Gene Details
Entrez Gene ID: | 19094 |
Gene Name: | mitogen-activated protein kinase 11 |
Gene Aliases: |
P38b, Prkm11, Sapk2, Sapk2b, p38-2, p38beta, p38beta2 |
Location: |
Chr.15:89142481-89149623 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk11 | NM_011161.5 | NP_035291.4 | ||
| AF135185.1 | AAD30116.1 | |||
| AK144846.1 | ||||
| AK161190.1 | ||||
| AK161562.1 | BAE36465.1 | |||
| BC057211.1 | AAH57211.1 | |||
| BC064737.1 | AAH64737.1 | |||
| BC092526.1 | AAH92526.1 |
More Information
Set Membership: |
|

Genomic Map