Assay Details
Target Gene Details
Entrez Gene ID: | 18521 |
Gene Name: | poly(rC) binding protein 2 |
Gene Aliases: |
AW412548, Hnrpx, alphaCP-2 |
Location: |
Chr.15:102470222-102500059 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcbp2 | NM_001103165.1 | NP_001096635.1 | ||
| NM_001103166.1 | NP_001096636.1 | |||
| NM_001174073.1 | NP_001167544.1 | |||
| NM_011042.2 | NP_035172.2 | |||
| AK012297.1 | ||||
| AK161527.1 | BAE36444.1 | |||
| AK169268.1 | BAE41029.1 | |||
| BQ900329.1 | ||||
| EU604548.1 | ||||
| L19661.1 | AAA03705.1 |
More Information
Set Membership: |
|

Genomic Map