Assay Details
Target Gene Details
Entrez Gene ID: | 77055 |
Gene Name: | keratin 76 |
Gene Aliases: |
2310001L23Rik |
Location: |
Chr.15:101884351-101892920 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt76 | NM_001033177.2 | NP_001028349.1 | ||
| AK009075.1 | ||||
| AK036802.1 | ||||
| AK137676.1 | BAE23456.1 |
More Information
Set Membership: |
|

Genomic Map