Assay Details
Target Gene Details
Entrez Gene ID: | 74775 |
Gene Name: | limb region 1 like |
Gene Aliases: |
1110013E13Rik, AA415653, D15Ertd735e, Limr |
Location: |
Chr.15:98903914-98918550 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lmbr1l | NM_029098.3 | NP_083374.1 | ||
| AK003656.1 | BAB22919.1 | |||
| AK030589.1 | BAC27033.1 | |||
| AK049110.1 | BAC33547.1 | |||
| AK143891.1 | BAE25586.1 | |||
| AK154968.1 | BAE32960.1 | |||
| AK170328.1 | BAE41722.1 | |||
| AK173121.1 | BAD32399.1 | |||
| AY052398.1 | AAL13076.1 | |||
| BC023397.1 | AAH23397.1 |
More Information
Set Membership: |
|

Genomic Map