Assay Details
Target Gene Details
Entrez Gene ID: | 29857 |
Gene Name: | mitogen-activated protein kinase 12 |
Gene Aliases: |
AW123708, Erk6, P38gamma, Prkm12, Sapk3 |
Location: |
Chr.15:89130584-89141593 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk12 | NM_013871.3 | NP_038899.1 | ||
| AK011286.1 | ||||
| BC002134.1 | 1 | 309 | AAH02134.1 | |
| BC021640.1 | AAH21640.1 | |||
| Y13439.1 | CAA73850.1 |
More Information
Set Membership: |
|

Genomic Map