Assay Details
Target Gene Details
Entrez Gene ID: | 75646 |
Gene Name: | retinoic acid induced 14 |
Gene Aliases: |
1700008J19Rik, 1700020L11Rik, Ankycorbin, Norpeg, mKIAA1334 |
Location: |
Chr.15:10568971-10719267 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rai14 | NM_001166408.1 | NP_001159880.1 | ||
| NM_030690.3 | NP_109615.1 | |||
| AF202315.1 | AAG24483.1 | |||
| AF274866.1 | AAG25937.1 | |||
| AK129333.1 | BAC98143.1 | |||
| AK160119.1 | ||||
| BC052458.1 | AAH52458.1 |

Genomic Map