Assay Details
Target Gene Details
Entrez Gene ID: | 332131 |
Gene Name: | keratin 78 |
Gene Aliases: |
2310030B04Rik, Kb40 |
Location: |
Chr.15:101946001-101996609 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt78 | NM_212487.4 | NP_997652.4 | ||
| AK146611.1 | ||||
| AY651827.1 | AAV85506.1 | |||
| BC119600.1 | ||||
| BC119601.1 | ||||
| BC127241.1 | ||||
| BC127618.1 |

Genomic Map